siRNA Details
| Gene Symbol: Ubqln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198623.2 | NP_941025.1 | 2 | 777 | ||
| GenBank mRNA | AK077072.1 | - | 2 | 779 | ||
| BC100419.1 | - | 2 | 754 | 30918401 | ||
| BC150726.1 | - | 1 | 704 | 9087637 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqln3| Gene Symbol: Ubqln3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198623.2 | NP_941025.1 | 2 | 777 | ||
| GenBank mRNA | AK077072.1 | - | 2 | 779 | ||
| BC100419.1 | - | 2 | 754 | 30918401 | ||
| BC150726.1 | - | 1 | 704 | 9087637 | ||