siRNA Details
| Gene Symbol: Ubqlnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198624.3 | NP_941026.2 | 1 | 1415 | ||
| GenBank mRNA | AK076645.1 | - | 1 | 1406 | ||
| BC113134.1 | - | 1 | 1346 | 40062178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqlnl| Gene Symbol: Ubqlnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198624.3 | NP_941026.2 | 1 | 1415 | ||
| GenBank mRNA | AK076645.1 | - | 1 | 1406 | ||
| BC113134.1 | - | 1 | 1346 | 40062178 | ||