siRNA Details
| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172907.3 | NP_766495.1 | 3 | 990 | ||
| GenBank mRNA | AK032948.1 | - | 3 | 951 | ||
| AK140277.1 | - | 3 | 743 | |||
| BC047207.1 | - | 3 | 977 | 6310165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfml1| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172907.3 | NP_766495.1 | 3 | 990 | ||
| GenBank mRNA | AK032948.1 | - | 3 | 951 | ||
| AK140277.1 | - | 3 | 743 | |||
| BC047207.1 | - | 3 | 977 | 6310165 | ||