siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 13 | 3216 | ||
| GenBank mRNA | AF539744.1 | - | 13 | 2972 | ||
| AK046675.1 | - | 12 | 3393 | |||
| BC145639.1 | - | 11 | 3041 | |||
| BC145640.1 | - | 12 | 3065 | |||

Cd163l1
Genomic Map