siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 7, 8 | 2230 | ||
| GenBank mRNA | AF539744.1 | - | 7, 8 | 1986 | ||
| AK046675.1 | - | 6, 7 | 2354 | |||
| AK088010.1 | - | 7, 8 | 2217 | |||
| BC145639.1 | - | 6, 7 | 2079 | |||
| BC145640.1 | - | 6, 7 | 2079 | |||

Cd163l1
Genomic Map