siRNA Details
| Gene Symbol: Cd163l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172909.4 | NP_766497.3 | 8, 9 | 2550 | ||
| GenBank mRNA | AF539744.1 | - | 8, 9 | 2306 | ||
| AK046675.1 | - | 7, 8 | 2674 | |||
| AK088010.1 | - | 8, 9 | 2537 | |||
| BC145639.1 | - | 7, 8 | 2399 | |||
| BC145640.1 | - | 7, 8 | 2399 | |||

Cd163l1
Genomic Map