siRNA Details
| Gene Symbol: Tnfrsf26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175649.5 | NP_783580.1 | 3 | 173 | ||
| GenBank mRNA | AK142511.1 | - | 234 | |||
| AK154666.1 | - | 3 | 284 | |||
| AY165628.1 | - | 3 | 173 | |||
| BC111108.1 | - | 3 | 161 | |||
| BC138819.1 | - | 3 | 222 | 8861841 | ||
| BC138820.1 | - | 3 | 223 | 8861842 | ||
| BC152357.1 | - | 3 | 169 | |||
| BC152358.1 | - | 3 | 173 | 40055748 | ||

Tnfrsf26
Genomic Map