siRNA Details
| Gene Symbol: Myo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081397.1 | NP_001074866.1 | 15 | 2016 | ||
| GenBank mRNA | AB429291.1 | - | 13 | 1624 | ||
| AK082350.1 | - | 15 | 1752 | |||
| BC072580.1 | - | 15 | 2028 | |||
| BC082311.1 | - | 15 | 1882 | |||
| BC151049.1 | - | 15 | 1970 | 9087962 | ||
| BC151051.1 | - | 15 | 1971 | 9087964 | ||
| BC157966.1 | - | 15 | 1971 | 9007290 | ||
| BC172122.1 | - | 15 | 1971 | 9054408 | ||

Myo16
Genomic Map