siRNA Details
| Gene Symbol: Lonrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081150.1 | NP_001074619.1 | 5 | 1542 | ||
| GenBank mRNA | BC094431.1 | - | 2 | 350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lonrf1| Gene Symbol: Lonrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081150.1 | NP_001074619.1 | 5 | 1542 | ||
| GenBank mRNA | BC094431.1 | - | 2 | 350 | ||