siRNA Details
| Gene Symbol: PRCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005973.4 | NP_005964.3 | 5, 6 | 1608 | ||
| XM_005245313.2 | XP_005245370.1 | 6, 7 | 1704 | |||
| XM_005245314.1 | XP_005245371.1 | 4, 5 | 1512 | |||
| GenBank mRNA | AK289872.1 | - | 5, 6 | 1548 | ||
| BC004913.2 | - | 5, 6 | 1524 | 3535413 | ||
| BC010450.1 | - | 5, 6 | 1536 | 4179105 | ||
| BU942864.1 | - | 2, 3 | 214 | |||
| KU178273.1 | - | 5, 6 | 1318 | |||
| KU178274.1 | - | 6, 7 | 1385 | |||
| KU178275.1 | - | 4, 5 | 1222 | |||
| X97124.1 | - | 5, 6 | 1508 | |||

PRCC
Genomic Map