siRNA Details
| Gene Symbol: Triml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177742.4 | NP_808410.1 | 5, 6 | 974 | ||
| GenBank mRNA | AK077092.1 | - | 5, 6 | 976 | ||
| BC141202.1 | - | 5, 6 | 953 | 9055814 | ||
| BC145531.1 | - | 5, 6 | 956 | 9054094 | ||

Triml1
Genomic Map