siRNA Details
| Gene Symbol: Wdr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172152.1 | NP_001165623.1 | 24 | 3187 | ||
| NM_028220.3 | NP_082496.3 | 25 | 3210 | |||
| GenBank mRNA | AK044241.1 | - | 24 | 3188 | ||
| BC028268.1 | - | 25 | 3221 | 5366098 | ||
| BC042703.1 | - | 3 | 330 | |||

Wdr17
Genomic Map