siRNA Details
| Gene Symbol: Wdr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172152.1 | NP_001165623.1 | 29 | 3759 | ||
| NM_028220.3 | NP_082496.3 | 30 | 3782 | |||
| GenBank mRNA | AK044241.1 | - | 29 | 3760 | ||
| BC028268.1 | - | 30 | 3793 | 5366098 | ||
| BC042703.1 | - | 8 | 898 | |||

Wdr17
Genomic Map