siRNA Details
| Gene Symbol: Podnl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013384.2 | NP_001013402.2 | 6, 7 | 768 | ||
| GenBank mRNA | AK077813.1 | - | 1, 2 | 917 | ||
| BC063764.1 | - | 6, 7 | 768 | 30288186 | ||
| DQ303463.1 | - | 6, 7 | 753 | |||

Podnl1
Genomic Map