siRNA Details
| Gene Symbol: Pskh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173432.2 | NP_775608.1 | 3 | 1196 | ||
| GenBank mRNA | AK053397.1 | - | 3 | 1230 | ||
| AK153861.1 | - | 3 | 1243 | |||
| AK154570.1 | - | 3 | 1226 | |||
| AK166338.1 | - | 3 | 1220 | |||
| AK182310.1 | - | 1 | 33 | |||
| AK191303.1 | - | 2 | 290 | |||
| AK219022.1 | - | 1 | 33 | |||
| BC050128.1 | - | 3 | 1196 | 30024345 | ||

Pskh1
Genomic Map