siRNA Details
| Gene Symbol: Sprtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111141.1 | NP_001104611.1 | 4 | 980 | ||
| GenBank mRNA | AK154380.1 | - | 4 | 983 | ||
| BC158088.1 | - | 4 | 835 | 9007378 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprtn| Gene Symbol: Sprtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111141.1 | NP_001104611.1 | 4 | 980 | ||
| GenBank mRNA | AK154380.1 | - | 4 | 983 | ||
| BC158088.1 | - | 4 | 835 | 9007378 | ||