siRNA Details
| Gene Symbol: Cwf19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027545.2 | NP_081821.1 | 8 | 1257 | ||
| GenBank mRNA | AK031423.1 | - | 8 | 1255 | ||
| AK045423.1 | - | 8 | 1258 | |||
| AK083386.1 | - | 8 | 1270 | |||
| BC043480.1 | - | 8 | 1224 | |||
| BC125483.1 | - | 8 | 1294 | 40129998 | ||
| BC125485.1 | - | 8 | 1294 | 40130000 | ||

Cwf19l2
Genomic Map