siRNA Details
| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170787.1 | NP_001164258.1 | 6 | 1045 | ||
| GenBank mRNA | AK032746.1 | - | 2 | 813 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cntn5| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170787.1 | NP_001164258.1 | 6 | 1045 | ||
| GenBank mRNA | AK032746.1 | - | 2 | 813 | ||