siRNA Details
| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033359.2 | NP_001028531.2 | 4 | 699 | ||
| NM_001170787.1 | NP_001164258.1 | 8 | 1363 | |||
| GenBank mRNA | AK138164.1 | - | 4 | 700 | ||
| AK162856.1 | - | 4 | 699 | |||

Cntn5
Genomic Map