siRNA Details
| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172920.4 | NP_766508.3 | 11 | 1169 | ||
| GenBank mRNA | AK143845.1 | - | 12 | 1174 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpy19l1| Gene Symbol: Dpy19l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172920.4 | NP_766508.3 | 11 | 1169 | ||
| GenBank mRNA | AK143845.1 | - | 12 | 1174 | ||