siRNA Details
| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033534.1 | NP_001028706.1 | 3 | 388 | ||
| GenBank mRNA | AK086930.1 | - | 3 | 349 | ||
| AK165695.1 | - | 3 | 342 | |||
| BC139816.1 | - | 360 | 40140447 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Layn| Gene Symbol: Layn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033534.1 | NP_001028706.1 | 3 | 388 | ||
| GenBank mRNA | AK086930.1 | - | 3 | 349 | ||
| AK165695.1 | - | 3 | 342 | |||
| BC139816.1 | - | 360 | 40140447 | |||