siRNA Details
| Gene Symbol: Snx14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172926.3 | NP_766514.2 | 8 | 794 | ||
| GenBank mRNA | AK046839.1 | - | 8 | 736 | ||
| AK049371.1 | - | 8 | 773 | |||
| AK082389.1 | - | 7 | 688 | |||
| BC080799.1 | - | 8 | 779 | 4948538 | ||

Snx14
Genomic Map