siRNA Details
| Gene Symbol: Gm4981 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034869.2 | NP_001030041.1 | 1 | 892 | ||
| GenBank mRNA | AK145460.1 | - | 1 | 893 | ||
| BC147818.1 | - | 1 | 696 | 9087535 | ||
| BC147822.1 | - | 1 | 696 | 9087539 | ||

Gm4981
Genomic Map