siRNA Details
| Gene Symbol: AA414768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272033.1 | NP_001258962.1 | 1 | 1157 | ||
| GenBank mRNA | BG086291.2 | - | 1 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AA414768| Gene Symbol: AA414768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272033.1 | NP_001258962.1 | 1 | 1157 | ||
| GenBank mRNA | BG086291.2 | - | 1 | 287 | ||