siRNA Details
| Gene Symbol: Gm4988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BB429596.2 | - | 1 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4988| Gene Symbol: Gm4988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BB429596.2 | - | 1 | 119 | ||