siRNA Details
| Gene Symbol: Slitrk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178740.4 | NP_848855.2 | 3 | 1934 | ||
| GenBank mRNA | AK032263.1 | - | 2 | 1824 | ||
| AK039763.1 | - | 3 | 1875 | |||
| AK046301.1 | - | 3 | 1805 | |||
| AK053065.1 | - | 3 | 1935 | |||
| BC117890.1 | - | 3 | 1799 | 40096585 | ||
| BC117891.1 | - | 2 | 1732 | 40096591 | ||

Slitrk4
Genomic Map