siRNA Details
| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153169.2 | NP_694809.1 | 1 | 447 | ||
| GenBank mRNA | BC036726.1 | - | 1 | 485 | 5368858 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pnma3| Gene Symbol: Pnma3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153169.2 | NP_694809.1 | 1 | 447 | ||
| GenBank mRNA | BC036726.1 | - | 1 | 485 | 5368858 | |