siRNA Details
| Gene Symbol: Pdzd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029868.2 | NP_001025039.1 | 8 | 1808 | ||
| NM_001290540.1 | NP_001277469.1 | 6 | 1463 | |||
| GenBank mRNA | AK046101.1 | - | 6 | 1466 | ||
| BC056462.1 | - | 8 | 1154 | 5703229 | ||

Pdzd4
Genomic Map