siRNA Details
| Gene Symbol: Trim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137967.1 | - | 1 | 469 | ||
| Gene Symbol: Trim12c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146007.1 | NP_001139479.1 | 8 | 1651 | ||
| NM_175677.4 | NP_783608.3 | 8 | 1532 | |||
| GenBank mRNA | AK033764.1 | - | 8 | 1651 | ||
| AK146437.1 | - | 8 | 1611 | |||
| BC038937.1 | - | 8 | 1532 | 2648191 | ||

Trim5
Genomic Map