siRNA Details
Gene Symbol: Zfp711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177747.3 | NP_808415.2 | 5, 6 | 1320 | ||
GenBank mRNA | CN525862.1 | - | 3, 4 | 430 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177747.3 | NP_808415.2 | 5, 6 | 1320 | ||
GenBank mRNA | CN525862.1 | - | 3, 4 | 430 |