siRNA Details
| Gene Symbol: Nxf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024141.4 | NP_001019312.1 | 8 | 955 | ||
| GenBank mRNA | AB178899.1 | - | 8 | 958 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxf3| Gene Symbol: Nxf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024141.4 | NP_001019312.1 | 8 | 955 | ||
| GenBank mRNA | AB178899.1 | - | 8 | 958 | ||