siRNA Details
| Gene Symbol: PRKAB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005399.4 | NP_005390.1 | 6 | 750 | ||
| NR_103870.1 | 4 | 462 | ||||
| NR_103871.1 | 5 | 583 | ||||
| XM_011509729.1 | XP_011508031.1 | 6 | 750 | |||
| XM_017001697.1 | XP_016857186.1 | 5 | 629 | |||
| GenBank mRNA | AJ224538.1 | - | 5 | 566 | ||
| AK294863.1 | - | 5 | 555 | |||
| AK304069.1 | - | 4 | 343 | |||
| AK310982.1 | - | 6 | 631 | |||
| AK316005.1 | - | 5 | 461 | |||
| AK316183.1 | - | 4 | 434 | |||
| AL037763.1 | - | 5 | 547 | |||
| BC053610.1 | - | 6 | 705 | 6091476 | ||
| BU166852.1 | - | 2 | 67 | |||
| KJ534919.1 | - | 5 | 566 | |||
| KJ535024.1 | - | 4 | 399 | |||
| KJ535025.1 | - | 5 | 566 | |||

PRKAB2
Genomic Map