siRNA Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172150.4 | NP_742162.2 | 13 | 1257 | ||
| GenBank mRNA | AK088467.1 | - | 13 | 1258 | ||
| AK144188.1 | - | 13 | 1357 | |||
| AK167194.1 | - | 13 | 1273 | |||
| BC037708.1 | - | 13 | 1226 | 4222723 | ||

Ift52
Genomic Map