siRNA Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172150.4 | NP_742162.2 | 8, 9 | 768 | ||
| GenBank mRNA | AK088467.1 | - | 8, 9 | 769 | ||
| AK144188.1 | - | 8, 9 | 868 | |||
| AK160745.1 | - | 8, 9 | 773 | |||
| AK167194.1 | - | 8, 9 | 784 | |||
| AK178241.1 | - | 2, 3 | 135 | |||
| BC037708.1 | - | 8, 9 | 737 | 4222723 | ||

Ift52
Genomic Map