siRNA Details
| Gene Symbol: Pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291211.1 | NP_001278140.1 | 5 | 824 | ||
| NM_153594.3 | NP_705822.1 | 5 | 824 | |||
| GenBank mRNA | AB182640.1 | - | 5 | 765 | ||
| AK031335.1 | - | 5 | 779 | |||
| AK079927.1 | - | 2 | 100 | |||
| AK088800.1 | - | 5 | 763 | |||
| AK146080.1 | - | 5 | 771 | |||
| BC031360.1 | - | 4 | 641 | 5359242 | ||
| BC040385.1 | - | 5 | 775 | 3499499 | ||

Pcmtd2
Genomic Map