siRNA Details
Gene Symbol: Defb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139223.3 | NP_631969.1 | 2 | 114 | ||
GenBank mRNA | AJ437649.1 | - | 2 | 114 | ||
AJ575427.1 | - | 2 | 85 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
