siRNA Details
| Gene Symbol: Atxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139227.4 | NP_631973.3 | 6 | 1148 | ||
| GenBank mRNA | AF455111.1 | - | 5 | 888 | ||
| AK046679.1 | - | 6 | 1149 | |||
| BC035459.1 | - | 5 | 801 | |||
| BC137846.1 | - | 6 | 1069 | 8860865 | ||

Atxn7
Genomic Map