siRNA Details
| Gene Symbol: Rhbdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139228.3 | NP_631974.1 | 9 | 1318 | ||
| GenBank mRNA | AJ313479.1 | - | 9 | 1318 | ||
| AK041678.1 | - | 6 | 896 | |||
| BC054784.1 | - | 9 | 1154 | 6817213 | ||
| BC056969.1 | - | 9 | 1154 | 6817213 | ||

Rhbdl3
Genomic Map