siRNA Details
| Gene Symbol: Fktn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139309.4 | NP_647470.1 | 1, 2 | 225 | ||
| GenBank mRNA | AB077383.1 | - | 2, 3 | 409 | ||
| AJ511807.1 | - | 2, 3 | 245 | |||
| AK085931.1 | - | 1, 2 | 226 | |||
| AK144743.1 | - | 2, 3 | 254 | |||

Fktn
Genomic Map