siRNA Details
| Gene Symbol: Bivm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144558.4 | NP_653141.3 | 2 | 782 | ||
| GenBank mRNA | AF411386.1 | - | 3 | 785 | ||
| AK032375.1 | - | 2 | 742 | |||
| AK034329.1 | - | 2 | 744 | |||
| AK047404.1 | - | 2 | 785 | |||
| BC082584.1 | - | 2 | 588 | 30612339 | ||

Bivm
Genomic Map