siRNA Details
| Gene Symbol: Bivm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144558.4 | NP_653141.3 | 6 | 1286 | ||
| GenBank mRNA | AF411386.1 | - | 7 | 1289 | ||
| AK032375.1 | - | 6 | 1246 | |||
| AK034329.1 | - | 6 | 1248 | |||
| BC082584.1 | - | 6 | 1092 | 30612339 | ||

Bivm
Genomic Map