siRNA Details
| Gene Symbol: Bivm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144558.4 | NP_653141.3 | 2 | 601 | ||
| GenBank mRNA | AF411386.1 | - | 3 | 604 | ||
| AK032375.1 | - | 2 | 561 | |||
| AK034329.1 | - | 2 | 563 | |||
| AK047404.1 | - | 2 | 604 | |||
| BC082584.1 | - | 2 | 407 | 30612339 | ||

Bivm
Genomic Map