siRNA Details
| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144942.4 | NP_659191.1 | 13 | 1370 | ||
| GenBank mRNA | AK005015.1 | - | 13 | 1371 | ||
| AY033912.1 | - | 13 | 1287 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csad| Gene Symbol: Csad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144942.4 | NP_659191.1 | 13 | 1370 | ||
| GenBank mRNA | AK005015.1 | - | 13 | 1371 | ||
| AY033912.1 | - | 13 | 1287 | |||