siRNA Details
| Gene Symbol: Cd207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144943.3 | NP_659192.2 | 4 | 733 | ||
| GenBank mRNA | AJ302711.1 | - | 4 | 733 | ||
| AK142731.1 | - | 4 | 675 | |||
| AY026050.1 | - | 4 | 675 | |||
| AY078415.1 | - | 4 | 675 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd207| Gene Symbol: Cd207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144943.3 | NP_659192.2 | 4 | 733 | ||
| GenBank mRNA | AJ302711.1 | - | 4 | 733 | ||
| AK142731.1 | - | 4 | 675 | |||
| AY026050.1 | - | 4 | 675 | |||
| AY078415.1 | - | 4 | 675 | |||