siRNA Details
| Gene Symbol: Klhl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178741.3 | NP_848856.1 | 3 | 831 | ||
| GenBank mRNA | AK031028.1 | - | 3 | 832 | ||
| AK034618.1 | - | 3 | 797 | |||
| AK049960.1 | - | 2 | 719 | |||
| AK184451.1 | - | 1 | 333 | |||
| AK212769.1 | - | 1 | 346 | |||
| AY101769.1 | - | 2 | 576 | |||
| BC086802.1 | - | 2 | 692 | 30612176 | ||

Klhl8
Genomic Map