siRNA Details
Gene Symbol: Prokr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144944.3 | NP_659193.3 | 3 | 808 | ||
GenBank mRNA | AF487279.1 | - | 2 | 486 | ||
AK030458.1 | - | 2 | 208 | |||
AK041586.1 | - | 3 | 732 | |||
AK080980.1 | - | 3 | 759 | |||
BC043116.1 | - | 3 | 761 | 6408655 | ||
BC057557.1 | - | 3 | 761 | 6408655 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
