siRNA Details
| Gene Symbol: Lgi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144945.2 | NP_659194.1 | 7 | 1124 | ||
| GenBank mRNA | AJ487515.1 | - | 7 | 1147 | ||
| AK122570.1 | - | 8 | 1065 | |||
| AY841361.1 | - | 8 | 925 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgi2| Gene Symbol: Lgi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144945.2 | NP_659194.1 | 7 | 1124 | ||
| GenBank mRNA | AJ487515.1 | - | 7 | 1147 | ||
| AK122570.1 | - | 8 | 1065 | |||
| AY841361.1 | - | 8 | 925 | |||