siRNA Details
| Gene Symbol: Slc25a28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145156.1 | NP_660138.1 | 2 | 564 | ||
| GenBank mRNA | AF377994.1 | - | 2 | 382 | ||
| AK155044.1 | - | 2 | 543 | |||
| BC025908.1 | - | 2 | 564 | 5043992 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a28| Gene Symbol: Slc25a28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145156.1 | NP_660138.1 | 2 | 564 | ||
| GenBank mRNA | AF377994.1 | - | 2 | 382 | ||
| AK155044.1 | - | 2 | 543 | |||
| BC025908.1 | - | 2 | 564 | 5043992 | ||