siRNA Details
| Gene Symbol: Slc25a28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145156.1 | NP_660138.1 | 4 | 1169 | ||
| GenBank mRNA | AF377993.1 | - | 3 | 638 | ||
| AF377994.1 | - | 4 | 987 | |||
| AK034173.1 | - | 1 | 2009 | |||
| AK155044.1 | - | 2 | 3362 | |||
| BC023172.1 | - | 2 | 680 | |||
| BC025908.1 | - | 4 | 1169 | 5043992 | ||

Slc25a28
Genomic Map