siRNA Details
| Gene Symbol: Rhobtb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153514.5 | NP_705734.4 | 3, 4 | 823 | ||
| GenBank mRNA | AF420001.1 | - | 2, 3 | 314 | ||
| AK037927.1 | - | 3, 4 | 504 | |||
| AK038196.1 | - | 3, 4 | 915 | |||
| AK081100.1 | - | 3, 4 | 752 | |||
| AK122354.1 | - | 3, 4 | 804 | |||
| AK171295.1 | - | 9, 10 | 1438 | |||
| BC026836.1 | - | 3, 4 | 804 | 4242008 | ||
| BC056954.1 | - | 3, 4 | 428 | 5685456 | ||

Rhobtb2
Genomic Map